Assay Details
Gene Symbol: QRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284543.2 | XP_015140029.1 | 3 - 4 | 326 | 71 | |
XM_419807.5 | XP_419807.2 | 3 - 4 | 384 | 71 | ||
GenBank mRNA | BX935823.2 | - | 3 - 4 | 347 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
