Assay Details
| Gene Symbol: SCN4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298254.2 | XP_015153740.1 | 3 - 4 | 609 | 75 | |
| XM_015298255.2 | XP_015153741.1 | 2 - 3 | 437 | 75 | ||
| XM_025143318.1 | XP_024999086.1 | 3 - 4 | 467 | 75 | ||
| XM_417913.5 | XP_417913.3 | 2 - 3 | 266 | 75 | ||
| GenBank mRNA | BX935954.2 | - | 1 - 2 | 163 | 75 | |
More Information
Set Membership: |
|

SCN4B
Genomic Map