Assay Details
| Gene Symbol: TBCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167729.1 | NP_001161201.1 | 1 - 2 | 101 | 126 | |
| GenBank mRNA | BX936008.2 | - | 1 - 2 | 149 | 126 | |
More Information
Set Membership: |
|

TBCA
Genomic Map