Assay Details
| Gene Symbol: TBCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167729.1 | NP_001161201.1 | 2 - 3 | 201 | 119 | |
| GenBank mRNA | BX936008.2 | - | 2 - 3 | 249 | 119 | |
More Information
Set Membership: |
|

TBCA
Genomic Map