Assay Details
| Gene Symbol: NUAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417962.6 | XP_417962.3 | 578 | 65 | ||
| GenBank mRNA | BX950299.1 | - | 558 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUAK2| Gene Symbol: NUAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417962.6 | XP_417962.3 | 578 | 65 | ||
| GenBank mRNA | BX950299.1 | - | 558 | 65 | ||
Set Membership: |
|