Assay Details
| Gene Symbol: GPR160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277472.1 | NP_001264401.1 | 1 | 1746 | 97 | |
| XM_015291665.2 | XP_015147151.1 | 2 | 2105 | 97 | ||
| GenBank mRNA | BX950382.1 | - | 1 | 1746 | 97 | |
More Information
Set Membership: |
|

GPR160
Genomic Map