Assay Details
| Gene Symbol: EGFL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277738.1 | NP_001264667.1 | 188 | 83 | ||
| GenBank mRNA | BX930768.1 | - | 122 | 83 | ||
| BX950717.1 | - | 188 | 83 | |||
More Information
Set Membership: |
|

EGFL6
Genomic Map