Assay Details
| Gene Symbol: PHYH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424238.6 | XP_424238.5 | 4 - 5 | 491 | 83 | |
| GenBank mRNA | CR353447.1 | - | 3 - 4 | 313 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PHYH| Gene Symbol: PHYH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424238.6 | XP_424238.5 | 4 - 5 | 491 | 83 | |
| GenBank mRNA | CR353447.1 | - | 3 - 4 | 313 | 83 | |
Set Membership: |
|