Assay Details
| Gene Symbol: HLCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015301496.2 | XP_015156982.1 | 3 - 4 | 460 | 90 | |
| XM_025145233.1 | XP_025001001.1 | 2 - 3 | 433 | 90 | ||
| XM_025145235.1 | XP_025001003.1 | 4 - 5 | 577 | 90 | ||
| XM_025145239.1 | XP_025001007.1 | 3 - 4 | 419 | 90 | ||
| XM_416725.6 | XP_416725.1 | 3 - 4 | 586 | 90 | ||
| GenBank mRNA | CR353737.1 | - | 3 - 4 | 511 | 90 | |
More Information
Set Membership: |
|

HLCS
Genomic Map