Assay Details
| Gene Symbol: ATP6AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252611.1 | NP_001239540.1 | 3 - 4 | 608 | 113 | |
| GenBank mRNA | CO422524.1 | - | 3 - 4 | 331 | 113 | |
| CR353783.1 | - | 3 - 4 | 324 | 113 | ||
More Information
Set Membership: |
|

ATP6AP1
Genomic Map