Assay Details
| Gene Symbol: RAB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640800.4 | XP_003640848.1 | 6 | 2690 | 117 | |
| GenBank mRNA | CR353886.1 | - | 1 | 450 | 117 | |
More Information
Set Membership: |
|

RAB12
Genomic Map