Assay Details
| Gene Symbol: ISY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414311.4 | XP_414311.1 | 8 - 9 | 620 | 71 | |
| GenBank mRNA | CR353909.1 | - | 7 - 8 | 554 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ISY1| Gene Symbol: ISY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414311.4 | XP_414311.1 | 8 - 9 | 620 | 71 | |
| GenBank mRNA | CR353909.1 | - | 7 - 8 | 554 | 71 | |
Set Membership: |
|