Assay Details
| Gene Symbol: SLC35F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935481.3 | XP_004935538.1 | 3 - 4 | 854 | 67 | |
| XM_015284407.2 | XP_015139893.1 | 2 - 3 | 505 | 67 | ||
| XM_419560.6 | XP_419560.3 | 3 - 4 | 794 | 67 | ||
| GenBank mRNA | BX929776.2 | - | 1 - 2 | 152 | 67 | |
More Information
Set Membership: |
|

SLC35F3
Genomic Map