Assay Details
| Gene Symbol: ZHX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025148151.1 | XP_025003919.1 | 2 | 1519 | 104 | |
| GenBank mRNA | BX931133.2 | - | 1 | 1047 | 104 | |
More Information
Set Membership: |
|

ZHX1
Genomic Map