Assay Details
| Gene Symbol: EMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198680.1 | NP_001185609.1 | 7 - 8 | 532 | 78 | |
| XM_015283011.2 | XP_015138497.1 | 5 - 6 | 435 | 78 | ||
| XM_025147115.1 | XP_025002883.1 | 7 - 8 | 531 | 78 | ||
| GenBank mRNA | BX931995.2 | - | 7 - 8 | 519 | 78 | |
| CN227744.1 | - | 7 - 8 | 503 | 78 | ||
More Information
Set Membership: |
|

EMC2
Genomic Map