Assay Details
| Gene Symbol: NSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415834.3 | XP_415834.1 | 5 - 6 | 539 | 74 | |
| GenBank mRNA | BX932125.2 | - | 5 - 6 | 758 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NSRP1| Gene Symbol: NSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415834.3 | XP_415834.1 | 5 - 6 | 539 | 74 | |
| GenBank mRNA | BX932125.2 | - | 5 - 6 | 758 | 74 | |
Set Membership: |
|