Assay Details
| Gene Symbol: FADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421073.6 | XP_421073.2 | 1 - 2 | 389 | 69 | |
| GenBank mRNA | BX932543.2 | - | 1 - 2 | 26 | 69 | |
| CR353855.1 | - | 1 - 2 | 375 | 69 | ||
More Information
Set Membership: |
|

FADD
Genomic Map