Assay Details
| Gene Symbol: SLC22A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146131.1 | NP_001139603.1 | 3 - 4 | 657 | 115 | |
| GenBank mRNA | BX932928.2 | - | 3 - 4 | 677 | 115 | |
| FN178490.1 | - | 3 - 4 | 657 | 115 | ||
More Information
Set Membership: |
|

SLC22A4
Genomic Map