Assay Details
| Gene Symbol: SERPINA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015287767.2 | XP_015143253.1 | 4 - 5 | 1294 | 87 | |
| XM_025150934.1 | XP_025006702.1 | 5 - 6 | 1352 | 87 | ||
| GenBank mRNA | BX933079.2 | - | 3 - 4 | 1192 | 87 | |
More Information
Set Membership: |
|

SERPINA10
Genomic Map