Assay Details
| Gene Symbol: C2H9ORF30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419077.6 | XP_419077.3 | 1 - 2 | 280 | 131 | |
| GenBank mRNA | BX933166.2 | - | 1 - 2 | 290 | 131 | |
| BX934590.1 | - | 1 - 2 | 288 | 131 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_419077.6 transcripts.
Set Membership: |
|

C2H9ORF30
Genomic Map