Assay Details
| Gene Symbol: UGT2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276390.2 | XP_015131876.1 | 3 | 1535 | 86 | |
| GenBank mRNA | BX950619.2 | - | 1 | 651 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UGT2A1| Gene Symbol: UGT2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276390.2 | XP_015131876.1 | 3 | 1535 | 86 | |
| GenBank mRNA | BX950619.2 | - | 1 | 651 | 86 | |
Set Membership: |
|