Assay Details
| Gene Symbol: USP51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294380.2 | XP_015149866.1 | 2 - 3 | 244 | 76 | |
| XM_015294383.2 | XP_015149869.1 | 2 - 3 | 244 | 76 | ||
| XM_025155364.1 | XP_025011132.1 | 3 - 4 | 380 | 76 | ||
| XM_025155366.1 | XP_025011134.1 | 2 - 3 | 244 | 76 | ||
| GenBank mRNA | CR386144.1 | - | 2 - 3 | 351 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015294380.2, XM_015294383.2, XM_025155364.1, XM_025155366.1 transcripts.
Set Membership: |
|

USP51
Genomic Map