Assay Details
| Gene Symbol: IL13RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420218.5 | XP_420218.3 | 10 | 92 | ||
| GenBank mRNA | CR386415.1 | - | 1 | 552 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL13RA1| Gene Symbol: IL13RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420218.5 | XP_420218.3 | 10 | 92 | ||
| GenBank mRNA | CR386415.1 | - | 1 | 552 | 92 | |
Set Membership: |
|