Assay Details
| Gene Symbol: PM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945285.3 | XP_004945342.2 | 6 - 7 | 580 | 103 | |
| XM_414903.6 | XP_414903.3 | 6 - 7 | 565 | 103 | ||
| GenBank mRNA | CR386453.1 | - | 6 - 7 | 438 | 103 | |
More Information
Set Membership: |
|

PM5
Genomic Map