Assay Details
| Gene Symbol: PM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945285.3 | XP_004945342.2 | 9 - 10 | 958 | 85 | |
| XM_414903.6 | XP_414903.3 | 9 - 10 | 943 | 85 | ||
| GenBank mRNA | CR386453.1 | - | 9 - 10 | 816 | 85 | |
More Information
Set Membership: |
|

PM5
Genomic Map