Assay Details
Gene Symbol: THBS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321549.1 | NP_001308478.1 | 20 - 21 | 2656 | 62 | |
XM_004937318.3 | XP_004937375.1 | 20 - 21 | 2727 | 62 | ||
GenBank mRNA | CR386821.1 | - | 6 - 7 | 515 | 62 | |
CR389535.1 | - | 18 - 19 | 2159 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
