Assay Details
| Gene Symbol: CCDC88B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941868.3 | XP_004941925.1 | 12 - 13 | 1417 | 71 | |
| XM_015287749.1 | XP_015143235.1 | 12 - 13 | 1336 | 71 | ||
| XM_025151373.1 | XP_025007141.1 | 12 - 13 | 1417 | 71 | ||
| GenBank mRNA | CR387222.1 | - | 2 - 3 | 245 | 71 | |
More Information
Set Membership: |
|

CCDC88B
Genomic Map