Assay Details
| Gene Symbol: RPL31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277755.1 | NP_001264684.1 | 4 - 5 | 393 | 79 | |
| GenBank mRNA | BX934620.1 | - | 1 - 2 | 568 | 79 | |
| CO635620.1 | - | 3 - 4 | 360 | 79 | ||
| CR387290.1 | - | 3 - 4 | 360 | 79 | ||
More Information
Set Membership: |
|

RPL31
Genomic Map