Assay Details
| Gene Symbol: DES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015290092.2 | XP_015145578.2 | 1 - 2 | 642 | 61 | |
| GenBank mRNA | AB011672.1 | - | 1 - 2 | 529 | 61 | |
More Information
Set Membership: |
|

DES
Genomic Map