Assay Details
| Gene Symbol: FZD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305216.1 | NP_001292145.1 | 1 | 1467 | 122 | |
| GenBank mRNA | AB029453.1 | - | 1 | 256 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FZD5| Gene Symbol: FZD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305216.1 | NP_001292145.1 | 1 | 1467 | 122 | |
| GenBank mRNA | AB029453.1 | - | 1 | 256 | 122 | |
Set Membership: |
|