Assay Details
| Gene Symbol: SDHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277398.1 | NP_001264327.1 | 7 - 8 | 930 | 80 | |
| GenBank mRNA | AF095939.1 | - | 3 - 4 | 396 | 80 | |
| BU298803.1 | - | 4 - 5 | 497 | 80 | ||
More Information
Set Membership: |
|

SDHA
Genomic Map