Assay Details
| Gene Symbol: HOXA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640697.4 | XP_003640745.1 | 2 | 980 | 139 | |
| GenBank mRNA | AY188521.1 | - | 1 | 148 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXA1| Gene Symbol: HOXA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640697.4 | XP_003640745.1 | 2 | 980 | 139 | |
| GenBank mRNA | AY188521.1 | - | 1 | 148 | 139 | |
Set Membership: |
|