Assay Details
| Gene Symbol: BCL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201487.2 | NP_001188416.2 | 1 - 2 | 117 | 67 | |
| GenBank mRNA | BU113828.1 | - | 1 - 2 | 49 | 67 | |
| BX929269.1 | - | 1 - 2 | 117 | 67 | ||
| HQ650869.1 | - | 1 - 2 | 75 | 67 | ||
More Information
Set Membership: |
|

BCL10
Genomic Map