Assay Details
| Gene Symbol: NDUFB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414844.5 | XP_414844.1 | 3 - 4 | 487 | 73 | |
| GenBank mRNA | BX929489.1 | - | 3 - 4 | 446 | 73 | |
More Information
Set Membership: |
|

NDUFB10
Genomic Map