Assay Details
| Gene Symbol: CMTM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025148025.1 | XP_025003793.1 | 1 - 2 | 167 | 90 | |
| GenBank mRNA | BX929722.1 | - | 1 - 2 | 134 | 90 | |
More Information
Set Membership: |
|

CMTM6
Genomic Map