Assay Details
| Gene Symbol: C2H9ORF30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419077.6 | XP_419077.3 | 2 - 3 | 717 | 118 | |
| GenBank mRNA | BX929920.1 | - | 2 - 3 | 663 | 118 | |
More Information
Set Membership: |
|

C2H9ORF30
Genomic Map