Assay Details
| Gene Symbol: MBIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421246.6 | XP_421246.2 | 2 - 3 | 605 | 70 | |
| GenBank mRNA | BX930446.1 | - | 2 - 3 | 282 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MBIP| Gene Symbol: MBIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421246.6 | XP_421246.2 | 2 - 3 | 605 | 70 | |
| GenBank mRNA | BX930446.1 | - | 2 - 3 | 282 | 70 | |
Set Membership: |
|