Assay Details
| Gene Symbol: COX7A2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277587.1 | NP_001264516.1 | 2 - 3 | 309 | 69 | |
| GenBank mRNA | BX932330.1 | - | 2 - 3 | 309 | 69 | |
| CK613988.1 | - | 2 - 3 | 243 | 69 | ||
| CN232533.1 | - | 2 - 3 | 256 | 69 | ||
More Information
Set Membership: |
|

COX7A2L
Genomic Map