Assay Details
| Gene Symbol: VTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284197.2 | XP_015139683.1 | 6 - 7 | 638 | 67 | |
| XM_419713.6 | XP_419713.4 | 5 - 6 | 576 | 67 | ||
| GenBank mRNA | BX932447.1 | - | 4 - 5 | 422 | 67 | |
More Information
Set Membership: |
|

VTA1
Genomic Map