Assay Details
| Gene Symbol: RBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277416.1 | NP_001264345.1 | 2 - 3 | 296 | 95 | |
| XM_015276907.2 | XP_015132393.1 | 2 - 3 | 2553 | 95 | ||
| GenBank mRNA | BX932569.1 | - | 2 - 3 | 296 | 95 | |
More Information
Set Membership: |
|

RBP1
Genomic Map