Assay Details
| Gene Symbol: TTC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278062.1 | NP_001264991.1 | 1 - 2 | 110 | 116 | |
| GenBank mRNA | BU239985.1 | - | 1 - 2 | 96 | 116 | |
| BX931970.2 | - | 1 - 2 | 128 | 116 | ||
| BX933530.1 | - | 1 - 2 | 93 | 116 | ||
More Information
Set Membership: |
|

TTC36
Genomic Map