Assay Details
| Gene Symbol: TTC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278062.1 | NP_001264991.1 | 2 - 3 | 302 | 131 | |
| XM_015297994.2 | XP_015153480.1 | 2 - 3 | 5058 | 131 | ||
| GenBank mRNA | BU239985.1 | - | 2 - 3 | 288 | 131 | |
| BX931970.2 | - | 2 - 3 | 320 | 131 | ||
| BX933530.1 | - | 2 - 3 | 285 | 131 | ||
More Information
Set Membership: |
|

TTC36
Genomic Map