Assay Details
| Gene Symbol: WARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421368.5 | XP_421368.1 | 9 - 10 | 1162 | 79 | |
| GenBank mRNA | BX933994.1 | - | 9 - 10 | 1144 | 79 | |
More Information
Set Membership: |
|

WARS
Genomic Map