Assay Details
| Gene Symbol: XIRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204348.3 | NP_989679.3 | 4 | 6007 | 77 | |
| XM_015281203.2 | XP_015136689.1 | 8 | 6651 | 77 | ||
| GenBank mRNA | D82816.2 | - | 1 | 352 | 77 | |
More Information
Set Membership: |
|

XIRP1
Genomic Map