Assay Details
| Gene Symbol: SRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205457.3 | NP_990788.3 | 2 - 3 | 317 | 109 | |
| XM_015296346.2 | XP_015151832.2 | 3 - 4 | 464 | 109 | ||
| XM_015296347.2 | XP_015151833.2 | 2 - 3 | 449 | 109 | ||
| XM_015296348.2 | XP_015151834.2 | 3 - 4 | 656 | 109 | ||
| GenBank mRNA | M57290.1 | - | 3 - 4 | 2384 | 109 | |
More Information
Set Membership: |
|

SRC
Genomic Map