Assay Details
| Gene Symbol: MAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937371.3 | XP_004937428.2 | 2 - 3 | 392 | 70 | |
| GenBank mRNA | X67778.1 | - | 318 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAP1B| Gene Symbol: MAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937371.3 | XP_004937428.2 | 2 - 3 | 392 | 70 | |
| GenBank mRNA | X67778.1 | - | 318 | 70 | ||
Set Membership: |
|