Assay Details
| Gene Symbol: PODXL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271894.1 | NP_001258823.1 | 4 - 5 | 1198 | 70 | |
| XM_015289480.2 | XP_015144966.1 | 4 - 5 | 1254 | 70 | ||
| XM_015289512.2 | XP_015144998.1 | 4 - 5 | 1254 | 70 | ||
| XM_015289559.2 | XP_015145045.1 | 4 - 5 | 1254 | 70 | ||
| GenBank mRNA | Y13978.1 | - | 1208 | 70 | ||
More Information
Set Membership: |
|

PODXL
Genomic Map