Assay Details
| Gene Symbol: KDELR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159715.1 | NP_001153187.1 | 4 - 5 | 721 | 82 | |
| GenBank mRNA | AJ719955.1 | - | 4 - 5 | 721 | 82 | |
More Information
Set Membership: |
|

KDELR2
Genomic Map