Assay Details
| Gene Symbol: ZBTB49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936168.3 | XP_004936225.1 | 3 - 4 | 1342 | 89 | |
| XM_015285814.1 | XP_015141300.1 | 5 - 6 | 1571 | 89 | ||
| XM_025149805.1 | XP_025005573.1 | 4 - 5 | 1500 | 89 | ||
| XM_025149806.1 | XP_025005574.1 | 4 - 5 | 1923 | 89 | ||
| GenBank mRNA | AJ720487.1 | - | 4 - 5 | 1254 | 89 | |
More Information
Set Membership: |
|

ZBTB49
Genomic Map