Assay Details
| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298107.2 | XP_015153593.1 | 7 - 8 | 561 | 111 | |
| XM_015298108.2 | XP_015153594.1 | 7 - 8 | 777 | 111 | ||
| XM_025143326.1 | XP_024999094.1 | 5 - 6 | 440 | 111 | ||
| XM_417860.5 | XP_417860.3 | 7 - 8 | 568 | 111 | ||
| GenBank mRNA | AJ866777.1 | - | 4 - 5 | 262 | 111 | |
| JX035870.1 | - | 4 - 5 | 291 | 111 | ||
More Information
Set Membership: |
|

C20orf173
Genomic Map